siRNA Details
| Gene Symbol: Stk32c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021302.3 | NP_067277.2 | 1 | 339 | ||
| GenBank mRNA | AB041542.1 | - | 1 | 319 | ||
| AK046439.1 | - | 1 | 317 | |||
| AU080917.1 | - | 339 | ||||
| BC026457.1 | - | 1 | 324 | 4481218 | ||

Stk32c
Genomic Map