siRNA Details
| Gene Symbol: Zfp112 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021307.2 | NP_067282.2 | 5 | 541 | ||
| GenBank mRNA | AF167319.1 | - | 4 | 521 | ||
| BC120886.2 | - | 5 | 537 | 40060889 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp112| Gene Symbol: Zfp112 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021307.2 | NP_067282.2 | 5 | 541 | ||
| GenBank mRNA | AF167319.1 | - | 4 | 521 | ||
| BC120886.2 | - | 5 | 537 | 40060889 | ||