siRNA Details
| Gene Symbol: Piwil1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021311.3 | NP_067286.1 | 6 | 677 | ||
| GenBank mRNA | AB032604.1 | - | 6 | 642 | ||
| AF438405.1 | - | 6 | 677 | |||
| BC066846.1 | - | 728 | ||||
| BC129857.1 | - | 5 | 527 | 40109791 | ||
| BC129858.1 | - | 5 | 527 | 40109793 | ||
| EF196092.1 | - | 4 | 487 | |||

Piwil1
Genomic Map