siRNA Details
| Gene Symbol: Rnf25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF171060.1 | - | 2 | 116 | ||
| AK002399.1 | - | 2 | 137 | |||
| AK171752.1 | - | 2 | 114 | |||
| BC022715.1 | - | 2 | 102 | |||
| BC037100.1 | - | 2 | 127 | |||
| BC048821.1 | - | 2 | 124 | 3966642 | ||
| BG081026.2 | - | 2 | 83 | |||

Rnf25
Genomic Map