siRNA Details
| Gene Symbol: Pglyrp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271476.1 | NP_001258405.1 | 3 | 453 | ||
| NM_001271477.1 | NP_001258406.1 | 2 | 345 | |||
| NM_001271478.1 | NP_001258407.1 | 3 | 453 | |||
| NM_001271479.1 | NP_001258408.1 | 2 | 345 | |||
| NM_021319.5 | NP_067294.2 | 2 | 345 | |||
| GenBank mRNA | AB084140.1 | - | 3 | 453 | ||
| AB084141.1 | - | 3 | 453 | |||
| AF149837.1 | - | 2 | 345 | |||
| AF149838.1 | - | 2 | 345 | |||
| AF315578.1 | - | 2 | 345 | |||
| AF315584.1 | - | 2 | 345 | |||
| AF317548.1 | - | 2 | 345 | |||
| AF392055.1 | - | 3 | 379 | |||
| AK039953.1 | - | 3 | 453 | |||
| AK050019.1 | - | 2 | 346 | |||
| AY282722.1 | - | 2 | 283 | |||
| BC019396.1 | - | 2 | 318 | 5124925 | ||
| EF204951.1 | - | 2 | 233 | |||

Pglyrp2
Genomic Map