siRNA Details
| Gene Symbol: Tnip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199275.2 | NP_001186204.1 | 3 | 400 | ||
| NM_001271456.1 | NP_001258385.1 | 3 | 295 | |||
| NM_021327.4 | NP_067302.2 | 2 | 242 | |||
| GenBank mRNA | AJ242778.1 | - | 2 | 214 | ||
| AK135338.1 | - | 2 | 221 | |||
| BC008186.1 | - | 2 | 159 | 3589311 | ||
| BC008665.1 | - | 3 | 266 | 3598934 | ||

Tnip1
Genomic Map