siRNA Details
| Gene Symbol: Bin3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021328.3 | NP_067303.1 | 3 | 129 | ||
| GenBank mRNA | AF244361.1 | - | 3 | 109 | ||
| AF271733.1 | - | 3 | 172 | |||
| AK005992.1 | - | 3 | 138 | |||
| AK040685.1 | - | 3 | 129 | |||
| AK170915.1 | - | 3 | 170 | |||
| AK210433.1 | - | 3 | 110 | |||
| BC026543.1 | - | 3 | 114 | 5040094 | ||

Bin3
Genomic Map