siRNA Details
| Gene Symbol: Rgr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021340.4 | NP_067315.1 | 3 | 362 | ||
| GenBank mRNA | AF076930.1 | - | 3 | 282 | ||
| AK044450.1 | - | 3 | 314 | |||
| AK044649.1 | - | 3 | 313 | |||
| AK149272.1 | - | 3 | 313 | |||
| BC082611.1 | - | 3 | 299 | |||
| BC086689.1 | - | 3 | 289 | |||
| BC119055.1 | - | 3 | 296 | 8733804 | ||
| CK622127.1 | - | 3 | 362 | |||
More Information
Set Membership: |
|

Rgr
Genomic Map