siRNA Details
Gene Symbol: Sorcs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252501.1 | NP_001239430.1 | 2, 3 | 812 | ||
NM_001290356.1 | NP_001277285.1 | 2, 3 | 812 | |||
NM_021377.3 | NP_067352.2 | 2, 3 | 812 | |||
GenBank mRNA | AF137367.1 | - | 2, 3 | 651 | ||
AF195056.1 | - | 2, 3 | 651 | |||
AF284755.1 | - | 2, 3 | 651 | |||
AK030440.1 | - | 2, 3 | 813 |

More Information
Other Species Targeted

Panther Classification:
