siRNA Details
| Gene Symbol: Stk32c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021302.3 | NP_067277.2 | 1, 2 | 397 | ||
| GenBank mRNA | AB041542.1 | - | 1, 2 | 377 | ||
| AK046439.1 | - | 1, 2 | 375 | |||
| AU080917.1 | - | 397 | ||||
| BC026457.1 | - | 1, 2 | 382 | 4481218 | ||

Stk32c
Genomic Map