siRNA Details
| Gene Symbol: Ntn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021320.3 | NP_067295.2 | 2 | 754 | ||
| GenBank mRNA | AF268066.1 | - | 2 | 349 | ||
| AF281278.1 | - | 2 | 602 | |||
| BB609851.1 | - | 2 | 625 | |||
| BC119182.1 | - | 2 | 436 | 8733931 | ||
| BC119184.1 | - | 2 | 436 | 8733933 | ||

Ntn4
Genomic Map