siRNA Details
| Gene Symbol: Bin3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021328.3 | NP_067303.1 | 4 | 181 | ||
| GenBank mRNA | AF244361.1 | - | 4 | 161 | ||
| AF271733.1 | - | 4 | 224 | |||
| AK005992.1 | - | 4 | 190 | |||
| AK040685.1 | - | 4 | 181 | |||
| AK170915.1 | - | 4 | 222 | |||
| AK210433.1 | - | 4 | 162 | |||
| BC026543.1 | - | 4 | 166 | 5040094 | ||

Bin3
Genomic Map