siRNA Details
| Gene Symbol: Rgr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301692.1 | NP_001288621.1 | 6 | 834 | ||
| NM_001301694.1 | NP_001288623.1 | 6 | 784 | |||
| NM_021340.4 | NP_067315.1 | 7 | 956 | |||
| GenBank mRNA | AF076930.1 | - | 7 | 876 | ||
| AK044450.1 | - | 7 | 908 | |||
| AK044649.1 | - | 7 | 907 | |||
| AK149272.1 | - | 7 | 907 | |||
| BC082611.1 | - | 7 | 893 | |||
| BC086689.1 | - | 6 | 1370 | |||
| BC119055.1 | - | 7 | 890 | 8733804 | ||
| BC144937.1 | - | 6 | 769 | |||
More Information
Set Membership: |
|

Rgr
Genomic Map