siRNA Details
| Gene Symbol: Tslp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021367.2 | NP_067342.1 | 5 | 358 | ||
| GenBank mRNA | AF232937.1 | - | 5 | 358 | ||
| AK087266.1 | - | 2 | 362 | |||
| BC130230.1 | - | 5 | 362 | 40141922 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tslp| Gene Symbol: Tslp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021367.2 | NP_067342.1 | 5 | 358 | ||
| GenBank mRNA | AF232937.1 | - | 5 | 358 | ||
| AK087266.1 | - | 2 | 362 | |||
| BC130230.1 | - | 5 | 362 | 40141922 | ||