siRNA Details
| Gene Symbol: Lancl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190984.1 | NP_001177913.1 | 10 | 2097 | ||
| NM_001190985.1 | NP_001177914.1 | 9 | 2315 | |||
| NM_021295.3 | NP_067270.1 | 10 | 2131 | |||
| GenBank mRNA | AJ294535.1 | - | 10 | 2124 | ||

Lancl1
Genomic Map