siRNA Details
| Gene Symbol: Rnf25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF171060.1 | - | 10 | 1151 | ||
| AK002399.1 | - | 10 | 1172 | |||
| AK171752.1 | - | 10 | 1149 | |||
| BC022715.1 | - | 10 | 1137 | |||
| BC037100.1 | - | 10 | 1159 | |||
| BC048821.1 | - | 10 | 1159 | 3966642 | ||
| BE853767.1 | - | 4 | 644 | |||
| BP774070.1 | - | 1 | 223 | |||

Rnf25
Genomic Map