siRNA Details
| Gene Symbol: Dnajb7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021317.2 | NP_067292.2 | 1 | 444 | ||
| GenBank mRNA | AB028855.1 | - | 1 | 444 | ||
| AK006201.1 | - | 1 | 470 | |||
| AK077186.1 | - | 1 | 473 | |||
| BC048738.1 | - | 1 | 433 | |||
| BC104396.1 | - | 1 | 391 | |||
| BC104397.1 | - | 1 | 391 | |||
| BC119478.2 | - | 1 | 391 | |||

Dnajb7
Genomic Map