siRNA Details
| Gene Symbol: Sh3kbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135727.2 | NP_001129199.1 | 14 | 1817 | ||
| NM_001135728.2 | NP_001129200.1 | 9 | 819 | |||
| NM_001290661.1 | NP_001277590.1 | 3 | 499 | |||
| NM_001290664.1 | NP_001277593.1 | 3 | 499 | |||
| NM_021389.6 | NP_067364.2 | 13 | 1382 | |||
| GenBank mRNA | AK004636.1 | - | 13 | 1382 | ||
| AK007283.1 | - | 3 | 486 | |||
| AK028299.1 | - | 9 | 823 | |||
| AK030839.1 | - | 14 | 1515 | |||
| AK049182.1 | - | 14 | 1818 | |||
| AK052260.1 | - | 3 | 174 | |||
| AK080930.1 | - | 14 | 1682 | |||
| AK142295.1 | - | 3 | 273 | |||
| AK161512.1 | - | 3 | 500 | |||
| AK161735.1 | - | 9 | 900 | |||
| AK172022.1 | - | 14 | 1798 | |||
| AK192695.1 | - | 2 | 33 | |||
| AK198973.1 | - | 2 | 33 | |||
| AK208632.1 | - | 3 | 191 | |||
| BC034646.1 | - | 13 | 1387 | |||
| BC052863.1 | - | 13 | 1327 | |||

Sh3kbp1
Genomic Map