siRNA Details
| Gene Symbol: Fam184b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021416.3 | NP_067391.3 | 1, 2 | 417 | ||
| GenBank mRNA | AB257856.1 | - | 1, 2 | 530 | ||
| AK081567.1 | - | 1, 2 | 201 | |||
| BC056379.1 | - | 1, 2 | 224 | 6850901 | ||
| CF539972.1 | - | 1, 2 | 417 | |||

Fam184b
Genomic Map