siRNA Details
| Gene Symbol: Fam181b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021427.2 | NP_067402.2 | 1 | 253 | ||
| GenBank mRNA | AB041569.1 | - | 1 | 213 | ||
| AK043957.1 | - | 1 | 268 | |||
| AK140685.1 | - | 1 | 205 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam181b| Gene Symbol: Fam181b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021427.2 | NP_067402.2 | 1 | 253 | ||
| GenBank mRNA | AB041569.1 | - | 1 | 213 | ||
| AK043957.1 | - | 1 | 268 | |||
| AK140685.1 | - | 1 | 205 | |||