siRNA Details
| Gene Symbol: Fzd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021458.2 | NP_067433.1 | 2 | 572 | ||
| GenBank mRNA | AK134261.1 | - | 2 | 508 | ||
| AK160554.1 | - | 2 | 531 | |||
| BC050965.1 | - | 2 | 533 | |||
| BC066186.1 | - | 2 | 500 | |||
| BC103714.1 | - | 2 | 500 | |||
| U43205.1 | - | 1 | 421 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_153474, AF323956.
Set Membership: |
|

Fzd3
Genomic Map