siRNA Details
Gene Symbol: Trem1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021406.5 | NP_067381.1 | 2 | 223 | ||
GenBank mRNA | AF241219.1 | - | 2 | 178 | ||
AK089439.1 | - | 2 | 192 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Trem1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021406.5 | NP_067381.1 | 2 | 223 | ||
GenBank mRNA | AF241219.1 | - | 2 | 178 | ||
AK089439.1 | - | 2 | 192 |