siRNA Details
| Gene Symbol: Rab37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021411.4 | NP_067386.3 | 2, 3 | 232 | ||
| GenBank mRNA | AB232633.1 | - | 2, 3 | 211 | ||
| AF233582.1 | - | 2, 3 | 211 | |||
| AK039716.1 | - | 2, 3 | 226 | |||
| AK040216.1 | - | 2, 3 | 232 | |||
| AK050483.1 | - | 2, 3 | 226 | |||
| BC050757.1 | - | 2, 3 | 232 | |||
| BC128020.1 | - | 2, 3 | 230 | 40136290 | ||
| BY131048.1 | - | 2, 3 | 243 | |||

Rab37
Genomic Map