siRNA Details
| Gene Symbol: Rnf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021419.1 | NP_067394.1 | 2 | 358 | ||
| GenBank mRNA | AJ242721.1 | - | 302 | |||
| AK076180.1 | - | 2 | 338 | |||
| AK147168.1 | - | 2 | 380 | |||
| AK219887.1 | - | 2 | 133 | |||
| BC021778.1 | - | 2 | 358 | 4239442 | ||
| CT010295.1 | - | 2 | 221 | |||

Rnf8
Genomic Map