siRNA Details
| Gene Symbol: Shank3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021423.3 | NP_067398.2 | 9 | 1007 | ||
| GenBank mRNA | AB231013.1 | - | 9 | 1007 | ||
| AJ245904.1 | - | 7 | 861 | |||
| KF612592.1 | - | 5 | 523 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Shank3| Gene Symbol: Shank3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021423.3 | NP_067398.2 | 9 | 1007 | ||
| GenBank mRNA | AB231013.1 | - | 9 | 1007 | ||
| AJ245904.1 | - | 7 | 861 | |||
| KF612592.1 | - | 5 | 523 | |||