siRNA Details
| Gene Symbol: Nkain4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021426.4 | NP_067401.3 | 5, 6 | 608 | ||
| GenBank mRNA | AB030182.1 | - | 5, 6 | 586 | ||
| AK005262.1 | - | 5, 6 | 579 | |||
| AK007632.1 | - | 5, 6 | 573 | |||
| AK021096.1 | - | 5, 6 | 603 | |||
| BC061107.1 | - | 5, 6 | 593 | 30295742 | ||
| BC089330.1 | - | 5, 6 | 560 | 30358178 | ||
| EF058055.1 | - | 5, 6 | 533 | |||

Nkain4
Genomic Map