siRNA Details
| Gene Symbol: Fzd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021458.2 | NP_067433.1 | 4 | 947 | ||
| GenBank mRNA | AK134261.1 | - | 4 | 883 | ||
| AK160554.1 | - | 4 | 906 | |||
| BC050965.1 | - | 4 | 908 | |||
| U43205.1 | - | 3 | 796 | |||
More Information
Set Membership: |
|

Fzd3
Genomic Map