siRNA Details
| Gene Symbol: Slco1c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177772.1 | NP_001171243.1 | 3 | 607 | ||
| NM_021471.2 | NP_067446.1 | 3 | 607 | |||
| GenBank mRNA | AK045782.1 | - | 610 | |||
| AK048926.1 | - | 3 | 577 | |||
| AK161010.1 | - | 577 | ||||
| AY007379.1 | - | 3 | 443 | |||
| BC078456.1 | - | 3 | 557 | 30360663 | ||

Slco1c1
Genomic Map