siRNA Details
| Gene Symbol: Ngly1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021504.3 | NP_067479.2 | 2, 3 | 307 | ||
| GenBank mRNA | AF250927.1 | - | 2, 3 | 294 | ||
| AK028248.1 | - | 2, 3 | 308 | |||
| AK148931.1 | - | 2, 3 | 310 | |||
| AK169205.1 | - | 2, 3 | 295 | |||
| BC028961.1 | - | 2, 3 | 261 | 5366023 | ||

Ngly1
Genomic Map