siRNA Details
| Gene Symbol: Hnrnph1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021510.2 | NP_067485.1 | 4 | 481 | ||
| GenBank mRNA | AK052983.1 | - | 4 | 435 | ||
| AK088169.1 | - | 4 | 434 | |||
| AK161491.1 | - | 579 | ||||
| AK178902.1 | - | 1 | 88 | |||
| BC016459.1 | - | 4 | 449 | |||
| BC027002.1 | - | 3 | 394 | |||
| BC042187.1 | - | 4 | 416 | 5376052 | ||
| BC056224.1 | - | 3 | 411 | 6515270 | ||
| BU058854.1 | - | 2 | 234 | |||
| Y14196.1 | - | 4 | 439 | |||

Hnrnph1
Genomic Map