siRNA Details
| Gene Symbol: Nup160 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021512.2 | NP_067487.1 | 3 | 853 | ||
| GenBank mRNA | AF104415.2 | - | 3 | 853 | ||
| AK155492.1 | - | 2 | 227 | |||
| AK160685.1 | - | 2 | 215 | |||
| AK164802.1 | - | 2 | 206 | |||
| AK171218.1 | - | 2 | 239 | |||
| BC052450.1 | - | 2 | 217 | 6409240 | ||
| BC054523.1 | - | 2 | 217 | 6409240 | ||

Nup160
Genomic Map