siRNA Details
| Gene Symbol: Trem1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021406.5 | NP_067381.1 | 4 | 1436 | ||
| GenBank mRNA | AK089439.1 | - | 4 | 1405 | ||
| AK171753.1 | - | 4 | 1405 | |||
| BC111875.1 | - | 3 | 1048 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trem1| Gene Symbol: Trem1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021406.5 | NP_067381.1 | 4 | 1436 | ||
| GenBank mRNA | AK089439.1 | - | 4 | 1405 | ||
| AK171753.1 | - | 4 | 1405 | |||
| BC111875.1 | - | 3 | 1048 | |||