siRNA Details
| Gene Symbol: Nkain4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021426.4 | NP_067401.3 | 6, 7 | 692 | ||
| GenBank mRNA | AB030182.1 | - | 6, 7 | 670 | ||
| AK005262.1 | - | 6, 7 | 663 | |||
| AK007632.1 | - | 6, 7 | 657 | |||
| AK021096.1 | - | 6, 7 | 687 | |||
| BC061107.1 | - | 6, 7 | 677 | 30295742 | ||
| BC089330.1 | - | 6, 7 | 644 | 30358178 | ||
| EF058055.1 | - | 617 | ||||

Nkain4
Genomic Map