siRNA Details
| Gene Symbol: Hs1bp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021429.3 | NP_067404.2 | 4 | 536 | ||
| GenBank mRNA | AJ132192.1 | - | 4 | 530 | ||
| AK040654.1 | - | 4 | 536 | |||
| AK084802.1 | - | 4 | 529 | |||
| AK154696.1 | - | 4 | 532 | |||
| AK170838.1 | - | 4 | 523 | |||
| BC095977.1 | - | 4 | 533 | |||

Hs1bp3
Genomic Map