siRNA Details
| Gene Symbol: 1700123O20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021437.2 | NP_067412.1 | 2 | 1391 | ||
| GenBank mRNA | AB041657.1 | - | 2 | 1346 | ||
| AK007261.1 | - | 2 | 792 | |||
| AK167989.1 | - | 2 | 575 | |||
| AK170890.1 | - | 2 | 724 | |||
| BC011283.1 | - | 2 | 520 | 3581864 | ||

1700123O20Rik
Genomic Map