siRNA Details
| Gene Symbol: CHRNA9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017581.3 | NP_060051.2 | 3 | 488 | ||
| GenBank mRNA | AF227732.1 | - | 3 | 331 | ||
| AJ243342.1 | - | 3 | 365 | |||
| BC113549.1 | - | 3 | 444 | 8322601 | ||
| BC113575.1 | - | 3 | 444 | 8322627 | ||

CHRNA9
Genomic Map