siRNA Details
| Gene Symbol: Maea | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021500.2 | NP_067475.2 | 5 | 615 | ||
| GenBank mRNA | AF263247.1 | - | 5 | 577 | ||
| AK007307.1 | - | 5 | 586 | |||
| AK049424.1 | - | 5 | 635 | |||
| AK075637.1 | - | 4 | 455 | |||
| AK088854.1 | - | 5 | 623 | |||
| AK145800.1 | - | 5 | 678 | |||
| AK152523.1 | - | 5 | 623 | |||
| AK168788.1 | - | 5 | 641 | |||
| BC039054.1 | - | 5 | 601 | 2648667 | ||
| BC058687.1 | - | 5 | 613 | 6411707 | ||

Maea
Genomic Map