siRNA Details
| Gene Symbol: Nup160 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021512.2 | NP_067487.1 | 27, 28 | 3740 | ||
| GenBank mRNA | AF104415.2 | - | 27, 28 | 3740 | ||
| AK172911.1 | - | 13, 14 | 1466 | |||
| BC052450.1 | - | 26, 27 | 3098 | 6409240 | ||
| BC054523.1 | - | 26, 27 | 3098 | 6409240 | ||

Nup160
Genomic Map