siRNA Details
| Gene Symbol: Rab2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021518.3 | NP_067493.1 | 4 | 422 | ||
| GenBank mRNA | AB232585.1 | - | 4 | 203 | ||
| AK042862.1 | - | 4 | 392 | |||
| AK079071.1 | - | 4 | 423 | |||
| AK135421.1 | - | 4 | 406 | |||
| AK146179.1 | - | 4 | 407 | |||
| AK150030.1 | - | 4 | 454 | |||
| AK159607.1 | - | 4 | 443 | |||
| AK160333.1 | - | 4 | 423 | |||
| AK163320.1 | - | 4 | 405 | |||
| AK180826.1 | - | 38 | ||||
| AK184879.1 | - | 3 | 106 | |||
| BC058382.1 | - | 4 | 420 | 6820798 | ||
| X95403.1 | - | 4 | 410 | |||

Rab2a
Genomic Map