siRNA Details
Gene Symbol: Jarid2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205043.1 | NP_001191972.1 | 2 | 723 | ||
NM_001205044.1 | NP_001191973.1 | 3 | 396 | |||
NM_021878.3 | NP_068678.1 | 3 | 442 | |||
GenBank mRNA | AK044888.1 | - | 2 | 627 | ||
AK045214.1 | - | 2 | 528 | |||
AK147226.1 | - | 2 | 723 | |||
BC026056.1 | - | 2 | 630 | |||
BC050129.1 | - | 2 | 644 | |||
BC051010.1 | - | 2 | 626 | |||
BC052444.1 | - | 3 | 313 | 6406875 | ||
BC060695.1 | - | 3 | 313 | 6406875 | ||
D31967.1 | - | 3 | 332 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
