siRNA Details
| Gene Symbol: Qk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159516.1 | NP_001152988.1 | 3 | 815 | ||
| NM_001159517.1 | NP_001152989.1 | 3 | 815 | |||
| NM_021881.2 | NP_068681.1 | 3 | 815 | |||
| GenBank mRNA | AF090402.1 | - | 4 | 975 | ||
| AF090403.1 | - | 4 | 975 | |||
| AF091047.1 | - | 975 | ||||
| AK005169.1 | - | 3 | 201 | |||
| AK132303.1 | - | 3 | 816 | |||
| BC053426.1 | - | 4 | 487 | 6335004 | ||
| BC056346.1 | - | 3 | 341 | 5705171 | ||
| U44940.1 | - | 3 | 834 | |||

Qk
Genomic Map