siRNA Details
| Gene Symbol: Ppp2r3c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021529.3 | NP_067504.2 | 3 | 371 | ||
| GenBank mRNA | AB041577.1 | - | 3 | 360 | ||
| AJ238247.2 | - | 2 | 198 | |||
| AK013403.1 | - | 2 | 167 | |||
| AK017550.1 | - | 2 | 154 | |||
| AK028063.1 | - | 3 | 336 | |||
| AK028254.1 | - | 2 | 290 | |||
| AK033405.1 | - | 3 | 369 | |||
| AK040589.1 | - | 2 | 199 | |||
| AK087572.1 | - | 2 | 194 | |||
| AK088240.1 | - | 2 | 280 | |||
| AK163192.1 | - | 370 | ||||
| BC024754.1 | - | 3 | 334 | 5375405 | ||
| BC138295.1 | - | 2 | 167 | 8861314 | ||
| BC138296.1 | - | 2 | 167 | 8861315 | ||

Ppp2r3c
Genomic Map