siRNA Details
| Gene Symbol: Wsb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021539.4 | NP_067514.2 | 3 | 357 | ||
| GenBank mRNA | AF033188.1 | - | 2 | 201 | ||
| AF072881.1 | - | 3 | 278 | |||
| AK136416.1 | - | 2 | 257 | |||
| AK157999.1 | - | 3 | 316 | |||
| AK202391.1 | - | 2 | 182 | |||
| BC055100.1 | - | 3 | 357 | 6589250 | ||

Wsb2
Genomic Map