siRNA Details
| Gene Symbol: Hgh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021555.2 | NP_067530.2 | 5 | 1014 | ||
| GenBank mRNA | AB041552.1 | - | 5 | 1014 | ||
| AK085815.1 | - | 5 | 1008 | |||
| BC061251.1 | - | 5 | 978 | 30249656 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hgh1| Gene Symbol: Hgh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021555.2 | NP_067530.2 | 5 | 1014 | ||
| GenBank mRNA | AB041552.1 | - | 5 | 1014 | ||
| AK085815.1 | - | 5 | 1008 | |||
| BC061251.1 | - | 5 | 978 | 30249656 | ||