siRNA Details
| Gene Symbol: Nek7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021605.3 | NP_067618.1 | 6 | 571 | ||
| GenBank mRNA | AF217650.1 | - | 5 | 422 | ||
| AK035502.1 | - | 6 | 570 | |||
| AK078639.1 | - | 6 | 580 | |||
| AK088173.1 | - | 6 | 620 | |||
| AK149308.1 | - | 6 | 619 | |||
| AK165583.1 | - | 6 | 549 | |||
| BC026541.1 | - | 6 | 563 | |||
| BC037697.1 | - | 6 | 548 | 3994871 | ||

Nek7
Genomic Map