siRNA Details
| Gene Symbol: TRPC7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167576.1 | NP_001161048.1 | 2 | 755 | ||
| NM_001167577.1 | NP_001161049.1 | 2 | 755 | |||
| NM_020389.2 | NP_065122.1 | 2 | 755 | |||
| GenBank mRNA | AJ272034.1 | - | 1 | 533 | ||
| AJ421783.1 | - | 1 | 533 | |||
| AJ549088.1 | - | 1 | 533 | |||
| AJ549089.1 | - | 1 | 533 | |||
| AJ549090.1 | - | 1 | 533 | |||
| AY167926.1 | - | 1 | 533 | |||
| AY167927.1 | - | 1 | 533 | |||
| BC128185.1 | - | 1 | 533 | 40113370 | ||
| BC128186.1 | - | 1 | 533 | |||

TRPC7
Genomic Map