siRNA Details
| Gene Symbol: Ap4s1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021710.3 | NP_068356.1 | 6 | 894 | ||
| GenBank mRNA | AF092093.1 | - | 6 | 823 | ||
| AK005283.1 | - | 6 | 644 | |||
| BC053339.1 | - | 6 | 654 | 5370721 | ||
| BP426557.1 | - | 4 | 349 | |||

Ap4s1
Genomic Map