siRNA Details
| Gene Symbol: Cenph | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021886.1 | NP_068686.1 | 4, 5 | 401 | ||
| GenBank mRNA | AB017634.1 | - | 4, 5 | 401 | ||
| AK006208.1 | - | 4, 5 | 385 | |||
| AK011264.1 | - | 4, 5 | 386 | |||
| AK011738.1 | - | 4, 5 | 386 | |||
| BC025084.1 | - | 4, 5 | 352 | 3501244 | ||

Cenph
Genomic Map