siRNA Details
| Gene Symbol: Cd274 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021893.3 | NP_068693.1 | 3 | 268 | ||
| GenBank mRNA | AF233517.1 | - | 2 | 201 | ||
| AF317088.1 | - | 2 | 185 | |||
| AK154403.1 | - | 3 | 247 | |||
| BC066841.1 | - | 3 | 251 | 30101931 | ||
| BY193530.1 | - | 3 | 264 | |||
| GQ904196.1 | - | 2 | 185 | |||

Cd274
Genomic Map