siRNA Details
| Gene Symbol: Stk25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021537.3 | NP_067512.3 | 12 | 1836 | ||
| GenBank mRNA | AF004934.1 | - | 11 | 1674 | ||
| AK005289.1 | - | 4 | 701 | |||
| AK148041.1 | - | 12 | 1810 | |||
| AK152917.1 | - | 12 | 1779 | |||
| AK211456.1 | - | 1 | 127 | |||
| BC029875.1 | - | 11 | 2554 | |||
| BC052913.1 | - | 12 | 1771 | 30057101 | ||
| BC071218.1 | - | 12 | 1831 | 4012695 | ||

Stk25
Genomic Map